siRNA Details
Gene Symbol: Cbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007624.3 | NP_031650.3 | 4 | 404 | ||
GenBank mRNA | AK002910.1 | - | 4 | 379 | ||
AK083957.1 | - | 4 | 319 | |||
AK146376.1 | - | 4 | 354 | |||
BC003840.1 | - | 4 | 330 | |||
BC018354.1 | - | 3 | 286 | 3990224 | ||
BC059831.1 | - | 4 | 307 | 5717629 | ||
BC108370.1 | - | 4 | 331 | 30728322 | ||
BC110376.1 | - | 4 | 361 | 3467208 | ||
CJ093936.1 | - | 4 | 405 | |||
X56683.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
