siRNA Details
Gene Symbol: Otogl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177567.1 | NP_001171038.1 | 33 | 3951 | ||
GenBank mRNA | GU140005.1 | - | 33 | 3952 |

More Information
Other Species Targeted

Gene Ontology Categories:
