siRNA Details
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183250.2 | NP_899073.1 | 4 | 620 | ||
GenBank mRNA | AK003762.1 | - | 4 | 621 | ||
AK169860.1 | - | 4 | 614 | |||
BC054363.1 | - | 4 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
