siRNA Details
Gene Symbol: Defb23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037933.2 | NP_001033022.1 | 2 | 229 | ||
GenBank mRNA | BC132568.1 | - | 2 | 229 | 40130845 | |
BC132570.1 | - | 2 | 229 | 40130847 | ||
DQ012034.1 | - | 2 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
