siRNA Details
Gene Symbol: Mroh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039557.4 | NP_001034646.2 | 21, 22 | 3014 | ||
GenBank mRNA | AK029507.1 | - | 21, 22 | 2986 | ||
AK132763.1 | - | 21, 22 | 2771 | |||
BC150937.1 | - | 21, 22 | 2738 | 9087848 |
