siRNA Details
Gene Symbol: Gm15501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CJ089270.1 | - | 1 | 328 | ||
DV661469.1 | - | 490 |
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009098.2 | NP_033124.1 | 4 | 375 | ||
GenBank mRNA | AK009023.1 | - | 4 | 328 | ||
AK010306.1 | - | 4 | 320 | |||
AK010650.1 | - | 4 | 375 | |||
AK011048.1 | - | 4 | 344 | |||
AK011058.1 | - | 4 | 328 | |||
AK011642.1 | - | 4 | 320 | |||
AK012435.1 | - | 4 | 328 | |||
AK012562.1 | - | 4 | 328 | |||
AK012665.1 | - | 4 | 347 | |||
AK019223.1 | - | 4 | 328 | |||
AK088660.1 | - | 4 | 328 | |||
AK145478.1 | - | 3 | 791 | |||
AK146303.1 | - | 4 | 328 | |||
AK146806.1 | - | 4 | 328 | |||
AK151544.1 | - | 4 | 328 | |||
AK166848.1 | - | 4 | 328 | |||
AK168609.1 | - | 4 | 328 | |||
BC027217.1 | - | 3 | 307 | 4205924 | ||
BC051446.1 | - | 3 | 324 | 30054488 | ||
BC081464.1 | - | 4 | 335 | |||
BC081465.1 | - | 3 | 307 | 6823252 | ||
BC100594.1 | - | 4 | 327 | 6750930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
