siRNA Details
Gene Symbol: Gm7008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004585.1 | - | 700 | |||
AK016351.1 | - | 796 | ||||
CB273562.1 | - | 1, 2 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
