siRNA Details
Gene Symbol: Krtap1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039502.2 | NP_001034591.1 | 1 | 1069 | ||
GenBank mRNA | AK131904.1 | - | 1 | 1070 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039502.2 | NP_001034591.1 | 1 | 1069 | ||
GenBank mRNA | AK131904.1 | - | 1 | 1070 |