siRNA Details
Gene Symbol: Krtap1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039502.2 | NP_001034591.1 | 1 | 1055 | ||
GenBank mRNA | AK131904.1 | - | 1 | 1056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
