siRNA Details
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110130.1 | NP_001103600.1 | 9 | 546 | ||
GenBank mRNA | AK008394.1 | - | 9 | 524 | ||
BY708305.1 | - | 9 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
