siRNA Details
Gene Symbol: Hbb-bh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127686.1 | NP_001121158.1 | 1, 2 | 191 | ||
GenBank mRNA | BC147125.1 | - | 1, 2 | 191 | 9056191 | |
BC147126.1 | - | 1, 2 | 191 | 9056176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
