siRNA Details
Gene Symbol: Slc25a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002283.1 | - | 2 | 577 | ||
AK003467.1 | - | 2 | 580 | |||
AK012750.1 | - | 2 | 582 | |||
AK012751.1 | - | 2 | 578 | |||
AK017881.1 | - | 2 | 575 | |||
AK088730.1 | - | 2 | 577 | |||
BC004570.1 | - | 2 | 560 | 3586336 | ||
BC086756.1 | - | 2 | 559 | 6818521 | ||
EF437333.1 | - | 1 | 85 | |||
U10404.1 | - | 2 | 500 | |||
U27316.1 | - | 2 | 585 | |||
X70847.1 | - | 2 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
