siRNA Details
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011482.4 | NP_035612.2 | 2 | 190 | ||
GenBank mRNA | AK004489.1 | - | 2 | 138 | ||
AK134943.1 | - | 128 | ||||
AK145391.1 | - | 2 | 192 | |||
AK145608.1 | - | 2 | 172 | |||
AK168808.1 | - | 2 | 160 | |||
AK168930.1 | - | 2 | 143 | |||
AK169103.1 | - | 2 | 174 | |||
BC026755.1 | - | 2 | 136 | 3154066 | ||
BC054450.1 | - | 2 | 136 | 6491298 | ||
BC083315.1 | - | 2 | 112 | 5714819 | ||
U95114.1 | - | 2 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
