siRNA Details
Gene Symbol: Ndufs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010888.2 | NP_035018.1 | 2 | 185 | ||
GenBank mRNA | AK147147.1 | - | 2 | 215 | ||
BC059730.1 | - | 2 | 158 | |||
BC076501.1 | - | 2 | 216 | |||
BC086933.1 | - | 2 | 158 | 6760537 | ||
BG081040.2 | - | 2 | 175 |

More Information
Other Species Targeted

Gene Ontology Categories:
