siRNA Details
Gene Symbol: Reck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016678.2 | NP_057887.2 | 9 | 954 | ||
GenBank mRNA | AK042266.1 | - | 9 | 956 | ||
AK143657.1 | - | 9 | 933 | |||
AK157885.1 | - | 9 | 942 | |||
BC053025.1 | - | 9 | 893 | |||
BC132197.1 | - | 9 | 982 | 40130474 | ||
BC138065.1 | - | 9 | 982 | 8861084 | ||
BC145133.1 | - | 9 | 983 |
