siRNA Details
Gene Symbol: Lce1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254760.1 | NP_001241689.1 | 2 | 334 | ||
GenBank mRNA | BY708617.1 | - | 2 | 334 |

UserName
Choose the brand aligned with your industry so we can best serve your needs.
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lce1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254760.1 | NP_001241689.1 | 2 | 334 | ||
GenBank mRNA | BY708617.1 | - | 2 | 334 |