siRNA Details
Gene Symbol: Gm12250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 375 | ||
GenBank mRNA | DQ508486.1 | - | 1 | 113 | ||
DQ508487.1 | - | 1 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
