siRNA Details
Gene Symbol: Gm7073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039240.3 | NP_001034329.2 | 4 | 969 | ||
GenBank mRNA | AK135660.1 | - | 4 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
