siRNA Details
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290501.1 | NP_001277430.1 | 9 | 3690 | ||
GenBank mRNA | BC068259.1 | - | 3 | 2930 | ||
BC151080.1 | - | 9 | 3564 | 9087993 | ||
BC151083.1 | - | 9 | 3564 | 9087996 |
