siRNA Details
Gene Symbol: Vmn1r169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166843.1 | NP_001160315.1 | 1 | 213 |
Gene Symbol: Vmn1r176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166721.1 | NP_001160193.1 | 1 | 213 |
