siRNA Details
Gene Symbol: H2afj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177688.4 | NP_808356.1 | 1 | 928 | ||
GenBank mRNA | AK053755.1 | - | 1 | 853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
