siRNA Details
Gene Symbol: Rpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009076.3 | NP_033102.2 | 5 | 404 | ||
GenBank mRNA | AK002973.1 | - | 5 | 403 | ||
AK008347.1 | - | 5 | 403 | |||
AK012349.1 | - | 5 | 403 | |||
AK012428.1 | - | 5 | 401 | |||
AK088464.1 | - | 5 | 403 | |||
AK144905.1 | - | 5 | 403 | |||
AK148566.1 | - | 1035 | ||||
AK150318.1 | - | 5 | 404 | |||
AK151532.1 | - | 5 | 403 | |||
AK151592.1 | - | 5 | 403 | |||
AK167761.1 | - | 5 | 423 | |||
AK168121.1 | - | 5 | 403 | |||
AK194205.1 | - | 3 | 164 | |||
AK198056.1 | - | 3 | 164 | |||
BC018321.1 | - | 5 | 400 | 3583073 | ||
BC075731.1 | - | 3 | 1022 | |||
BC081469.1 | - | 5 | 387 | 6824411 | ||
BC090393.1 | - | 382 | 5011867 | |||
L04280.1 | - | 5 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
