siRNA Details
Gene Symbol: Rpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048057.1 | NP_001041522.1 | 3, 4 | 185 | ||
NM_001048058.1 | NP_001041523.1 | 2, 3 | 111 | |||
NM_023372.2 | NP_075861.1 | 3, 4 | 168 | |||
GenBank mRNA | AB037665.1 | - | 3, 4 | 187 | ||
AK002659.1 | - | 3, 4 | 135 | |||
AK010277.1 | - | 3, 4 | 122 | |||
AK010518.2 | - | 3, 4 | 121 | |||
AK012391.1 | - | 3, 4 | 121 | |||
AK151517.1 | - | 3, 4 | 122 | |||
BC034339.1 | - | 3, 4 | 124 | 3985781 | ||
BC047046.1 | - | 2, 3 | 111 | |||
BC055346.1 | - | 3, 4 | 120 | 5003663 | ||
BC081447.1 | - | 2, 3 | 85 | |||
BC094258.1 | - | 2, 3 | 116 | 1245753 | ||
BC094644.1 | - | 2, 3 | 117 | 3328874 | ||
BU921494.1 | - | 3, 4 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
