siRNA Details
Gene Symbol: CK137956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134733.1 | NP_001128205.1 | 1 | 201 | ||
GenBank mRNA | BC147529.1 | - | 1 | 201 | 9087439 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CK137956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134733.1 | NP_001128205.1 | 1 | 201 | ||
GenBank mRNA | BC147529.1 | - | 1 | 201 | 9087439 |