siRNA Details
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288578.1 | NP_001275507.1 | 8 | 1096 | ||
NM_007410.3 | NP_031436.2 | 8 | 1114 | |||
GenBank mRNA | AK002790.1 | - | 8 | 1036 | ||
AK076507.1 | - | 8 | 1043 | |||
AK146949.1 | - | 8 | 1058 | |||
AK159803.1 | - | 8 | 1094 | |||
BC062879.1 | - | 8 | 1040 | 5691327 | ||
BC090978.1 | - | 8 | 1037 | 30313517 | ||
M84147.1 | - | 8 | 1064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
