siRNA Details
Gene Symbol: Ccdc94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028381.3 | NP_082657.1 | 3, 4 | 322 | ||
GenBank mRNA | AK013533.1 | - | 3, 4 | 336 | ||
AK168621.1 | - | 3, 4 | 411 | |||
BC031400.1 | - | 3, 4 | 322 | 4952601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
