siRNA Details
Gene Symbol: Vmn2r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104620.1 | NP_001098090.1 | 6 | 2500 |
Gene Symbol: Vmn2r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104622.1 | NP_001098092.1 | 6 | 2539 |
Gene Symbol: Vmn2r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104621.1 | NP_001098091.1 | 6 | 2500 |
