siRNA Details
Gene Symbol: Speer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027650.3 | NP_081926.2 | 2 | 229 | ||
GenBank mRNA | AK016676.1 | - | 2 | 216 | ||
CJ042389.1 | - | 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
