siRNA Details
Gene Symbol: Gm7168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122977.1 | NP_001116449.1 | 1 | 669 | ||
GenBank mRNA | BC107382.2 | - | 1 | 669 | 40057241 | |
BC125014.1 | - | 1 | 669 | 40057242 | ||
BC125267.1 | - | 1 | 669 | 40086171 |
