siRNA Details
Gene Symbol: Gp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163014.1 | NP_001156486.1 | 3 | 269 | ||
GenBank mRNA | AK079424.1 | - | 3 | 246 | ||
BC138170.1 | - | 3 | 253 | 8861189 | ||
BC145172.1 | - | 3 | 253 | 9053723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
