siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040696.1 | NP_001035786.1 | 10 | 3098 | ||
NM_001162414.1 | NP_001155886.1 | 9 | 3070 | |||
GenBank mRNA | BC141354.1 | - | 9 | 2999 | 9055966 | |
DQ117591.1 | - | 10 | 3098 | |||
DQ117592.1 | - | 9 | 2971 | |||
DQ117593.1 | - | 9 | 2971 | |||
DQ153214.1 | - | 10 | 3258 | |||
EF158039.1 | - | 9 | 3070 |
