siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040696.1 | NP_001035786.1 | 3 | 1070 | ||
GenBank mRNA | BC141354.1 | - | 2 | 971 | 9055966 | |
DQ117591.1 | - | 3 | 1070 | |||
DQ117592.1 | - | 2 | 943 | |||
DQ117593.1 | - | 2 | 943 | |||
DQ153214.1 | - | 3 | 1070 |
