siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040696.1 | NP_001035786.1 | 10 | 2985 | ||
NM_001162414.1 | NP_001155886.1 | 9 | 2957 | |||
GenBank mRNA | BC141354.1 | - | 9 | 2886 | 9055966 | |
DQ117583.1 | - | 3225 | ||||
DQ117584.1 | - | 8 | 3122 | |||
DQ117585.1 | - | 8 | 3035 | |||
DQ117586.1 | - | 8 | 3035 | |||
DQ117587.1 | - | 8 | 3035 | |||
DQ117588.1 | - | 8 | 3035 | |||
DQ117589.1 | - | 8 | 3035 | |||
DQ117590.1 | - | 8 | 3035 | |||
DQ117591.1 | - | 10 | 2985 | |||
DQ117592.1 | - | 9 | 2858 | |||
DQ117593.1 | - | 9 | 2858 | |||
DQ117597.1 | - | 8 | 2847 | |||
DQ117598.1 | - | 8 | 2847 | |||
DQ117599.1 | - | 8 | 2847 | |||
DQ117600.1 | - | 8 | 2852 | |||
DQ153214.1 | - | 10 | 3145 | |||
DQ860103.1 | - | 8 | 2852 | |||
DQ860104.1 | - | 8 | 2852 | |||
EF158039.1 | - | 9 | 2957 |
