siRNA Details
Gene Symbol: Vmn2r73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105186.1 | NP_001098656.1 | 3 | 818 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn2r73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105186.1 | NP_001098656.1 | 3 | 818 |