siRNA Details
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145423.2 | NP_663398.2 | 2 | 681 | ||
GenBank mRNA | AK039927.1 | - | 2 | 510 | ||
AK136457.1 | - | 2 | 693 | |||
AY484428.1 | - | 2 | 681 | |||
BC017691.1 | - | 636 | 4239161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
