siRNA Details
Gene Symbol: Aff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290814.1 | NP_001277743.1 | 19 | 3385 | ||
GenBank mRNA | BC052061.1 | - | 19 | 2715 | ||
BQ443842.1 | - | 3 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
