siRNA Details
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009096.3 | NP_033122.1 | 3 | 267 | ||
GenBank mRNA | AK012284.1 | - | 2 | 237 | ||
AK012828.1 | - | 3 | 268 | |||
AK013333.1 | - | 2 | 238 | |||
AK013825.1 | - | 267 | ||||
AK028205.1 | - | 3 | 270 | |||
AK050609.1 | - | 3 | 268 | |||
AK146952.1 | - | 3 | 268 | |||
AK161991.1 | - | 3 | 270 | |||
AK166680.1 | - | 3 | 269 | |||
AK166727.1 | - | 3 | 313 | |||
AK168050.1 | - | 3 | 268 | |||
AK168496.1 | - | 3 | 247 | |||
AK169624.1 | - | 3 | 266 | |||
BC010604.1 | - | 3 | 253 | 3481640 | ||
BC090392.1 | - | 3 | 254 | 4459055 | ||
BC092050.1 | - | 3 | 254 | 3666815 | ||
Y00348.1 | - | 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
