siRNA Details
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134138.1 | NP_598899.1 | 7 | 981 | ||
GenBank mRNA | AB031387.1 | - | 7 | 981 | ||
AK077569.1 | - | 7 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
