siRNA Details
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159697.1 | NP_001153169.1 | 8 | 1794 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159697.1 | NP_001153169.1 | 8 | 1794 |