siRNA Details
Gene Symbol: Gm7265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087684.1 | - | 1 | 1947 | ||
BC147147.1 | - | 1 | 194 | 9056180 | ||
BC147148.1 | - | 1 | 194 | 9056177 |

More Information
Other Species Targeted

