siRNA Details
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030754.4 | NP_110381.2 | 4 | 327 | ||
GenBank mRNA | BC020795.1 | - | 4 | 288 | 4767650 | |
M23699.1 | - | 3 | 251 | |||
M23700.1 | - | 3 | 251 | |||
M26152.1 | - | 4 | 274 | |||
X51440.1 | - | 2 | 37 | |||
X51445.1 | - | 3 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
