siRNA Details
Gene Symbol: Skint8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100466.1 | NP_001093936.2 | 4 | 601 | ||
GenBank mRNA | EF494903.1 | - | 4 | 601 | ||
EF494904.1 | - | 4 | 601 | |||
EU099307.1 | - | 4 | 601 |
