siRNA Details
Gene Symbol: Pin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027181.1 | NP_081457.1 | 4 | 406 | ||
GenBank mRNA | AK010338.1 | - | 4 | 375 | ||
BC119355.1 | - | 4 | 406 | 8734104 | ||
BC119357.1 | - | 4 | 406 | 8734106 |
Gene Symbol: Dnajb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135329.1 | - | 1 | 128 |

More Information
Other Species Targeted

Gene Ontology Categories:
