siRNA Details
Gene Symbol: Klra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101620.1 | NP_001095090.1 | 2 | 234 | ||
NM_010650.3 | NP_034780.1 | 171 | ||||
GenBank mRNA | AK143236.1 | - | 172 | |||
L78253.1 | - | 89 | ||||
U12889.1 | - | 2 | 285 |
Gene Symbol: Klra14-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY860969.1 | - | 89 | |||
AY860974.1 | - | 1 | 68 |
Gene Symbol: Klra21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053151.1 | NP_444381.1 | 201 | |||
GenBank mRNA | AF288380.1 | - | 201 | |||
AY338977.1 | - | 158 | ||||
AY338978.1 | - | 136 | ||||
AY860976.1 | - | 1 | 107 | |||
AY971806.1 | - | 95 | ||||
BC064073.1 | - | 197 | 874161 | |||
BC116399.1 | - | 178 | 40101978 | |||
BC116400.1 | - | 179 | 40101979 | |||
FJ159529.1 | - | 89 |
Gene Symbol: Klra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC141217.1 | - | 185 | 9055829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
