siRNA Details
Gene Symbol: Gm8994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142734.1 | NP_001136206.1 | 2 | 1514 | ||
GenBank mRNA | AK139713.1 | - | 2 | 1516 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm8994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142734.1 | NP_001136206.1 | 2 | 1514 | ||
GenBank mRNA | AK139713.1 | - | 2 | 1516 |