siRNA Details
Gene Symbol: Defb26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039120.2 | NP_001034209.1 | 2 | 311 | ||
GenBank mRNA | BC139131.1 | - | 2 | 311 | 8862153 | |
DQ012037.1 | - | 2 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
