siRNA Details
Gene Symbol: Angptl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039554.3 | NP_001034643.1 | 4 | 842 | ||
GenBank mRNA | AK031675.1 | - | 4 | 842 | ||
BC023373.1 | - | 4 | 829 | 5043159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
