siRNA Details
Gene Symbol: Smco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039558.2 | NP_001034647.2 | 2 | 626 | ||
GenBank mRNA | AK045581.1 | - | 2 | 603 | ||
AK047766.1 | - | 2 | 795 | |||
AK078719.1 | - | 2 | 630 | |||
BB624054.1 | - | 2 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
