siRNA Details
Gene Symbol: Sft2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134114.2 | NP_598875.2 | 4 | 417 | ||
GenBank mRNA | AK017471.1 | - | 2 | 161 | ||
AK151338.1 | - | 4 | 348 | |||
AK171776.1 | - | 2 | 182 | |||
BC006777.1 | - | 4 | 372 | |||
BC091770.1 | - | 4 | 418 | 4974098 | ||
BC130232.1 | - | 4 | 341 | 40141936 |

More Information
Other Species Targeted

Gene Ontology Categories:
