siRNA Details
Gene Symbol: Sft2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134114.2 | NP_598875.2 | 3 | 307 | ||
GenBank mRNA | AK151338.1 | - | 3 | 238 | ||
BC006777.1 | - | 3 | 262 | |||
BC091770.1 | - | 3 | 308 | 4974098 | ||
BC130232.1 | - | 3 | 231 | 40141936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
